missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidobenzoyl chloride, 95%

Product Code. p-7047267
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11384995

Brand: Thermo Scientific Alfa Aesar L18432.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16331-48-9 | C9H8ClNO2 | 197.618 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16331-48-9
Molecular Formula C9H8ClNO2
Molecular Weight (g/mol) 197.618
MDL Number MFCD02094022
InChI Key KRKXGCJUNKZXOY-UHFFFAOYSA-N
PubChem CID 7010341
IUPAC Name 4-acetamidobenzoyl chloride
SMILES CC(=O)NC1=CC=C(C=C1)C(=O)Cl

Specifications

Melting Point ∽145°C (decomposition)
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Synonym 4-acetylamino benzoyl chloride, benzoyl chloride,4-acetylamino, benzoyl chloride, 4-acetylamino, acmc-20amqa, 4-acetamidobenzoylchloride, 4-acetylaminobenzoyl chloride
Formula Weight 197.62
Percent Purity 95%
Chemical Name or Material 4-Acetamidobenzoyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.