missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetoxy-N-methyl-N-ethyltryptamine Fumarate (4-AcO-MET Fumarate; 4-Acetoxy-MET Fumarate) 1.0 mg/ml in Dimethyl Sulfoxide (as free base), LoGiCal
Description
4-Acetoxy-N-methyl-N-ethyltryptamine Fumarate (4-AcO-MET Fumarate; 4-Acetoxy-MET Fumarate) 1.0 mg/ml in Dimethyl Sulfoxide (as free base)
Specifications
Specifications
| Chemical Name or Material | 4-Acetoxy-N-methyl-N-ethyltryptamine Fumarate |
| CAS | 2711006-96-9 |
| Analyte or Component Names | 4-Acetoxy-N-methyl-N-ethyltryptamine Fumarate |
| Shipping Condition | Ice Pack (-20°C) |
| Molecular Formula | C15 H20 N2 O2 . C4 H4 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 4-Acetoxy-N-methyl-N-ethyltryptamine Fumarate, 4-AcO-MET Fumarate, 4-Acetoxy-MET Fumarate, 3-[2-(Ethylmethylamino)ethyl]-1 H-indol-4-ol 4-acetate fumarate, 1 H-Indol-4-ol, 3-[2-(ethylmethylamino)ethyl]-, 4-acetate, (2 E)-2-butenedioate (1:1) (ACI) |
| InChI Formula | InChI=1 S/C15H20N2O2.C4H4O4/c1-4-17(3)9-8-12-10-16-13-6-5-7-14(15(12)13)19-11(2)18;5-3(6)1-2-4(7)8/h5-7,10,16 H,4,8-9H2,1-3H3;1-2 H,(H,5,6)(H,7,8)/b;2-1+ |
| SMILES | CCN(C)CCc1c[nH]c2cccc(OC(=O)C)c12.OC(=O)\C=C\C(=O)O |
| IUPAC Name | (E)-but-2-enedioic acid;[3-[2-[ethyl(methyl)amino]ethyl]-1 H-indol-4-yl] acetate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?