missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxybenzoic Acid, Mikromol™

Product Code. 30050172
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050172 25 mg 25mg
30051060 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050172 Supplier Mikromol Supplier No. MM0133.110025
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Acetoxybenzoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Acetoxybenzoic Acid
CAS 2345-34-8
Analyte or Component Names 4-Acetoxybenzoic Acid
Shipping Condition Room Temperature
Molecular Formula C9 H8 O4
Certifications/Compliance ISO 17025
Synonym 4-(Acetyloxy)benzoic Acid, Benzoic acid, p-hydroxy-, acetate (6CI,7CI,8CI), 4-(Acetyloxy)benzoic acid, 4-Acetoxybenzoic acid, 4-Carboxyphenyl acetate, NSC 11340, p-Acetoxybenzoic acid, p-Acetyloxybenzoic acid, p-Carboxyphenyl acetate, p-Hydroxybenzoic acid acetate
InChI Formula InChI=1S/C9H8O4/c1-6(10)13-8-4-2-7(3-5-8)9(11)12/h2-5H,1H3,(H,11,12)
SMILES CC(=O)Oc1ccc(cc1)C(=O)O
IUPAC Name 4-acetyloxybenzoic acid
Molecular Weight (g/mol) 180.16
Quantity 25 mg
Formula Weight 180.0423 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.