missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-(trifluoromethyl)benzoic acid, 97+%

Product Code. 15405377
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15405377

Brand: Thermo Scientific Alfa Aesar H32626.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 393-06-6 | C8H6F3NO2 | 205.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 393-06-6
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.14
MDL Number MFCD03407959
InChI Key AMVHEVZYTGHASE-UHFFFAOYSA-N
Synonym 4-amino-2-trifluoromethyl benzoic acid, 5-amino-2-carboxybenzotrifluoride, 2-trifluoromethyl-4-aminobenzoic acid, pubchem1360, 4-amimo-2-trifluoromethyl benzoicacid, acmc-209j4m, ksc495q5d, rarechem al bo 0438, buttpark 44\01-93, 4-amino-2-trifluoromethyl benzoicacid
PubChem CID 3836325
IUPAC Name 4-amino-2-(trifluoromethyl)benzoic acid
SMILES NC1=CC(=C(C=C1)C(O)=O)C(F)(F)F

Specifications

Melting Point 183°C to 186°C
Quantity 1 g
Formula Weight 205.14
Percent Purity ≥97%
Chemical Name or Material 4-Amino-2-(trifluoromethyl)benzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.