missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-(trifluoromethoxy)benzoic acid, 95%, Thermo Scientific™

Product Code. p-4510628
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 10029511

Brand: Thermo Scientific Maybridge TL00679DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175278-22-5
Molecular Formula C8H6F3NO3
Molecular Weight (g/mol) 221.135
InChI Key IXJFWBNYFTWBOR-UHFFFAOYSA-N
Synonym 4-amino-3-trifluoromethoxy benzoic acid, rarechem al bo 0798, buttpark 83\07-32, 4-amino-3-trifluoromethoxy-benzoic acid, 4-amine-3-trifluoromethoxy benzoic acid, benzoic acid, 4-amino-3-trifluoromethoxy, 4-amino-3-trifluoromethoxybenzoicacid, pubchem14042, acmc-209e9s
PubChem CID 2735951
IUPAC Name 4-amino-3-(trifluoromethoxy)benzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)OC(F)(F)F)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 221.13
Percent Purity 95%
Chemical Name or Material 4-Amino-3-(trifluoromethoxy)benzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.