missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzoic Acid, LoGiCal

Product Code. 30464028
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464028 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464028 Supplier LoGiCal Supplier No. LGCFOR0094.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vitamin H1 (4-Aminobenzoic acid)
CAS 150-13-0
Analyte or Component Names Vitamin H1 (4-Aminobenzoic acid)
Shipping Condition Room Temperature
Molecular Formula C7 H7 N O2
Certifications/Compliance ISO 9001
Synonym Benzylpenicillin Procaine Imp. A (EP), Oxybuprocaine Hydrochloride Imp. A (EP), Oxybuprocaine Imp. A (EP), Tetracaine Hydrochloride Imp. A (EP), Tetracaine Imp. A (EP), 4-Aminobenzoic Acid, 4-Carboxyaniline, 4-Carboxyphenylamine, Actipol, Amben, Aniline-4-carboxylic acid, Anti-Chromotrichia factor, Anticanitic vitamin, Anticantic vitamin, Antichromotrichia factor, Bacterial vitamin H1, Chromotrichia factor, Hachemina, NSC 7627, PAB, PABA, Pabacyd, Pabafilm, Pabamine, Paraminol, Paranate, Romavit, Sunbrella, Trichochromogenic factor, Vitamin BX, Vitamin H', p-Aminobenzoic acid, p-Carboxyaniline, p-Carboxyphenylamine, Benzocaine Impurity G, Benzylpenicillin Procaine Impurity A, Oxybuprocaine Hydrochloride Impurity A, Tetracaine Hydrochloride Impurity A, Oxybuprocaine Impurity A, Tetracaine Impurity A
InChI Formula InChI=1S/C7H7NO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,8H2,(H,9,10)
Solution Type Neat
SMILES Nc1ccc(cc1)C(=O)O
IUPAC Name 4-aminobenzoic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 137.14
Quantity 10 mg
Formula Weight 137.0477 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.