missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoylbiphenyl, 99%

Product Code. 11423047
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423047 25 g 25g
11433047 100 g 100g
11443047 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423047 Supplier Thermo Scientific Alfa Aesar Supplier No. A15573.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2128-93-0 | C19H14O | 258.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2128-93-0
Molecular Formula C19H14O
Molecular Weight (g/mol) 258.32
MDL Number MFCD00003079
InChI Key LYXOWKPVTCPORE-UHFFFAOYSA-N
Synonym 4-benzoylbiphenyl, 4-phenylbenzophenone, p-phenylbenzophenone, benzophenone, 4-phenyl, p-benzoylbiphenyl, 4-diphenyl phenyl ketone, p-biphenylyl phenyl ketone, trigonal 12, 4-biphenylyl phenyl ketone, methanone, 1,1'-biphenyl-4-ylphenyl
PubChem CID 75040
IUPAC Name phenyl-(4-phenylphenyl)methanone
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C3=CC=CC=C3

Specifications

Melting Point 98°C to 103°C
Quantity 25 g
Beilstein 1876092
Formula Weight 258.32
Percent Purity 99%
Chemical Name or Material 4-Benzoylbiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.