missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzylaminocarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 11432290
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432290 1 g 1g
11442290 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432290 Supplier Thermo Scientific Alfa Aesar Supplier No. H29271.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073353-57-7
Molecular Formula C20H24BNO3
Molecular Weight (g/mol) 337.23
MDL Number MFCD09266184
InChI Key VAFYISMQZLEYKR-UHFFFAOYSA-N
Synonym n-benzyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 4-n-benzylaminocarbonyl phenylboronic acid, pinacol ester, 4-benzylaminocarbonyl benzeneboronic acid pinacol ester, 4-benzylamino-1-carbonyl phenyl boronic acid pinacol ester, amtb052, 4-n-benzylaminocarbonyl phenylboronic acid pinacol ester, n-benzyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide
PubChem CID 46739041
IUPAC Name N-benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(=O)NCC1=CC=CC=C1

Specifications

Melting Point 109°C to 113°C
Quantity 1 g
Formula Weight 337.22
Percent Purity 97%
Chemical Name or Material 4-(Benzylaminocarbonyl)benzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.