missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-amino)benzeneboronic acid, 97%

Product Code. 11402160
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11402160 1 g 1g
11412160 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11402160 Supplier Thermo Scientific Alfa Aesar Supplier No. H28799.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 380430-49-9 | C11H16BNO4 | 237.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380430-49-9
Molecular Formula C11H16BNO4
Molecular Weight (g/mol) 237.06
MDL Number MFCD02093054
InChI Key UBVOLHQIEQVXGM-UHFFFAOYSA-N
Synonym 4-boc-aminophenyl boronic acid, 4-n-boc-amino phenylboronic acid, 4-tert-butoxycarbonyl amino phenyl boronic acid, 4-tert-butoxycarbonylamino phenylboronic acid, 4-tert-butoxycarbonyl aminophenylboronic acid, 4-boc-amino benzeneboronic acid, 4-aminobenzeneboronic acid, n-boc protected, 4-tert-butoxycarbonyl amino phenylboronic acid, 4-2-methylpropan-2-yl oxycarbonylamino phenyl boronic acid, 4-tert-butoxycarbonylamino phenyl boronic acid
PubChem CID 3613184
IUPAC Name [4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)B(O)O

Specifications

Melting Point ∽200°C (decomposition)
Quantity 1 g
Formula Weight 237.06
Percent Purity 97%
Chemical Name or Material 4-(Boc-amino)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.