missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluoro-6-nitrophenol, 98%

Product Code. p-7031175
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15484837

Brand: Thermo Scientific Alfa Aesar H32153.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 320-76-3 | C6H3BrFNO3 | 236.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 320-76-3
Molecular Formula C6H3BrFNO3
Molecular Weight (g/mol) 236.00
MDL Number MFCD00013372
InChI Key ACQVEWFMUBXEMR-UHFFFAOYSA-N
Synonym 4-bromo-2-fluoro-6-nitrophenol, synquest 4654-b-46, phenol, 4-bromo-2-fluoro-6-nitro, 4-bromo-2-fluoro-6-nitro-phenol, pubchem2842, acmc-209xa8, 4-bromo-2-fluoro 6-nitrophenol, 4-bromo-6-fluoro-2-nitrophenol, 2-fluoro-4-bromo-6-nitro-phenol, phenol,4-bromo-2-fluoro-6-nitro
PubChem CID 688116
IUPAC Name 4-bromo-2-fluoro-6-nitrophenol
SMILES OC1=C(F)C=C(Br)C=C1[N+]([O-])=O

Specifications

Quantity 1 g
Formula Weight 236
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-fluoro-6-nitrophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.