missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylphenol, 97%

Product Code. 15405027
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15405027 25 g 25g
15485017 1 g 1g
15495017 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15405027 Supplier Thermo Scientific Alfa Aesar Supplier No. H32307.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2362-12-1 | C7H7BrO | 187.04 g/mol

4-Bromo-2-methylphenol, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-methylphenol, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2362-12-1
MDL Number MFCD00055435

Specifications

Melting Point 62°C to 66°C
Odor Phenol-like
Molecular Formula C13H9F3O
Quantity 25 g
Synonym 4-trifluoromethyl diphenyl ether, 1-phenoxy-4-trifluoromethyl benzene, 4-phenoxy-1-trifluoromethyl benzene, 4-phenoxybenzotrifluoride, trifluoromethyldiphenyl ether, 4-trifluoromethyl diphenylether, 4-phenoxy-alpha,alpha,alpha-trifluorotoluene
Solubility Information Slightly soluble in water.
InChI Key UZJUDUZMPNCXPF-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)C(F)(F)F
IUPAC Name 1-phenoxy-4-(trifluoromethyl)benzene
Molecular Weight (g/mol) 238.21
PubChem CID 2777517
Formula Weight 187.04
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methylphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.