missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylphenyl isocyanate, 98%, Thermo Scientific™

Product Code. 11357546
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357546 1 g 1g
11367546 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357546 Supplier Thermo Scientific Alfa Aesar Supplier No. L11095.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1591-98-6
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.046
MDL Number MFCD00037039
InChI Key NXAYRHQYINQHRF-UHFFFAOYSA-N
Synonym 4-bromo-2-methylphenyl isocyanate, benzene,4-bromo-1-isocyanato-2-methyl, 4-bromo-2-methylphenylisocyanate, 4-bromo-2-methylbenzenisocyanate, acmc-1bpqh, 5-bromo-2-isocyanatotoluene, 2-methyl-4-bromophenyl isocyanate, 4-bromanyl-1-isocyanato-2-methyl-benzene
PubChem CID 2735594
IUPAC Name 4-bromo-1-isocyanato-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Br)N=C=O

Specifications

Melting Point 47°C to 51°C
Flash Point >110°C (230°F)
Odor Irritating
Quantity 1 g
UN Number UN2811
Beilstein 7633645
Sensitivity Moisture sensitive
Formula Weight 212.05
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-methylphenyl isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.