missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzonitrile, 98%

Product Code. 10417290 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10417290

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-00-7 | C7H4BrN | 182.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-00-7
Molecular Formula C7H4BrN
Molecular Weight (g/mol) 182.02
MDL Number MFCD00001811
InChI Key HQSCPPCMBMFJJN-UHFFFAOYSA-N
Synonym p-bromobenzonitrile, benzonitrile, 4-bromo, benzonitrile, p-bromo, 1-bromo-4-cyanobenzene, 4-bromo benzonitrile, 4-cyano-bromobenzene, 4-bromo-4'-cyanobenzene, 4-cyanobromobenzene, bromobenzonitrile-4, 4-bromocyanobenzene
PubChem CID 12162
IUPAC Name 4-bromobenzonitrile
SMILES BrC1=CC=C(C=C1)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 111.0°C to 114.0°C
Color White to Yellow
Boiling Point 235.0°C to 237.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula BrC6H4CN
Quantity 10 g
Beilstein 09,354
Solubility Information Solubility in water: 0,2g/L. Other solubilities: soluble in acetone
Formula Weight 182.02
Percent Purity 98%
Physical Form Powder or Flakes
Chemical Name or Material 4-Bromobenzonitrile, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromobenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.