missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-5-sulfamoyl-2-[[((2RS)-tetrahydrofuran-2-yl)methyl]amino]benzoic Acid, Mikromol™
Description
4-Chloro-5-sulfamoyl-2-[[((2RS)-tetrahydrofuran-2-yl)methyl]amino]benzoic Acid
Specifications
Specifications
| Chemical Name or Material | Tetrahydro Furosemide |
| CAS | 4793-38-8 |
| Analyte or Component Names | Tetrahydro Furosemide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H15 Cl N2 O5 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | 5-(Aminosulfonyl)-4-chloro-2-[[(tetrahydro-2-furanyl)methyl]amino]benzoic acid, Anthranilic acid, 4-chloro-5-sulfamoyl-N-(tetrahydrofurfuryl)- (7CI,8CI), Furosemide Imp. F (EP), Furosemide Impurity F |
| InChI Formula | InChI=1S/C12H15ClN2O5S/c13-9-5-10(15-6-7-2-1-3-20-7)8(12(16)17)4-11(9)21(14,18)19/h4-5,7,15H,1-3,6H2,(H,16,17)(H2,14,18,19) |
| SMILES | NS(=O)(=O)c1cc(C(=O)O)c(NCC2CCCO2)cc1Cl |
| IUPAC Name | 4-chloro-2-(oxolan-2-ylmethylamino)-5-sulfamoylbenzoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?