missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline, Mikromol™

Product Code. 30052450
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052450 25 mg 25mg
30050714 100 mg 100mg
30460155 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30052450 Supplier Mikromol Supplier No. MM3297.010025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline
CAS 183322-18-1
Analyte or Component Names 4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline
Shipping Condition Room Temperature
Molecular Formula C14 H17 Cl N2 O4
Certifications/Compliance ISO 17025
Synonym 4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline, 4-Chloro-6,7-di(2-methoxyethoxy)quinazoline, 6,7-Bis(2-methoxyethoxy)-4-chloroquinazoline
InChI Formula InChI=1S/C14H17ClN2O4/c1-18-3-5-20-12-7-10-11(16-9-17-14(10)15)8-13(12)21-6-4-19-2/h7-9H,3-6H2,1-2H3
SMILES COCCOc1cc2ncnc(Cl)c2cc1OCCOC
IUPAC Name 4-chloro-6,7-bis(2-methoxyethoxy)quinazoline
Molecular Weight (g/mol) 312.75
Quantity 25 mg
Formula Weight 312.0877 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.