missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzaldehyde, Mikromol™

Product Code. 18021310
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18021310 25 mg 25mg
18002910 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18021310 Supplier Mikromol Supplier No. MM0017.030025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Chlorobenzaldehyde
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chlorobenzaldehyde
CAS 104-88-1
Analyte or Component Names 4-Chlorobenzaldehyde
Shipping Condition Room Temperature
Molecular Formula C7 H5 Cl O
Certifications/Compliance ISO 17025
Synonym 4-Chlorobenzaldehyde
InChI Formula InChI=1S/C7H5ClO/c8-7-3-1-6(5-9)2-4-7/h1-5H
SMILES Clc1ccc(C=O)cc1
IUPAC Name 4-chlorobenzaldehyde
Molecular Weight (g/mol) 140.57
Quantity 25 mg
Formula Weight 140.0029 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.