missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzamide, Mikromol™

Product Code. 17931906
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 17931906

Brand: Mikromol MM0250.060025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Chlorobenzamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chlorobenzamide
CAS 619-56-7
Analyte or Component Names 4-Chlorobenzamide
Shipping Condition Room Temperature
Molecular Formula C7 H6 Cl N O
Certifications/Compliance ISO 17025
Synonym Benzamide, 4-chloro-, Benzamide, p-chloro- (6CI,7CI,8CI), 4-Chlorobenzamide, NSC 406894, NSC 74687, p-Chlorobenzamide, p-Chlorobenzoic acid amide
InChI Formula InChI=1S/C7H6ClNO/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H2,9,10)
SMILES NC(=O)c1ccc(Cl)cc1
IUPAC Name 4-chlorobenzamide
Molecular Weight (g/mol) 155.58
Quantity 25 mg
Formula Weight 155.0138 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.