missing translation for 'onlineSavingsMsg'
Learn More

4'-Cyanoacetanilide, 98%

Product Code. p-7022256
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11442777

Brand: Thermo Scientific Alfa Aesar A14941.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35704-19-9 | C9H8N2O | 160.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35704-19-9
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.18
MDL Number MFCD00001814
InChI Key UFKRTEWFEYWIHD-UHFFFAOYSA-N
Synonym n-4-cyanophenyl acetamide, 4-cyanoacetanilide, 4-acetamidobenzonitrile, acetamide, n-4-cyanophenyl, 4'-cyanoacetanilide, p-acetamidobenzonitirle, acetanilide, 4'-cyano, p-acetylamino-benzonitrile, p-cyanoacetanilide, p-acetaminobenzonitrile
PubChem CID 37256
IUPAC Name N-(4-cyanophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)C#N

Specifications

Melting Point 204°C to 206°C
Quantity 5 g
Formula Weight 160.18
Percent Purity 98%
Chemical Name or Material 4'-Cyanoacetanilide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.