missing translation for 'onlineSavingsMsg'
Learn More

4'-Demethylepipodophyllotoxin, Mikromol™

Product Code. 30049521
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049521 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049521 Supplier Mikromol Supplier No. MM0492.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4'-Demethylepipodophyllotoxin
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4'-Demethylepipodophyllotoxin
CAS 6559-91-7
Analyte or Component Names 4'-Demethylepipodophyllotoxin
Shipping Condition Ice pack (-20°C)
Molecular Formula C21 H20 O8
Certifications/Compliance ISO 9001
Synonym 4'-Demethylepipodophyllotoxin, Teniposide Related Compound A, Etoposide Impurity E, Podophyllotoxin Impurity B
InChI Formula InChI=1S/C21H20O8/c1-25-15-3-9(4-16(26-2)20(15)23)17-10-5-13-14(29-8-28-13)6-11(10)19(22)12-7-27-21(24)18(12)17/h3-6,12,17-19,22-23H,7-8H2,1-2H3/t12-,17+,18-,19+/m0/s1
SMILES COc1cc(cc(OC)c1O)[C@H]2[C@@H]3[C@H](COC3=O)[C@H](O)c4cc5OCOc5cc24
IUPAC Name (5S,5aR,8aR,9R)-5-hydroxy-9-(4-hydroxy-3,5-dimethoxyphenyl)-5a,6,8a,9-tetrahydro-5H-[2]benzofuro[5,6-f][1,3]benzodioxol-8-one
Molecular Weight (g/mol) 400.38
Quantity 25 mg
Formula Weight 400.1158 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.