missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenol, 99%

Product Code. 11453944
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453944 25 g 25g
11463944 100 g 100g
11473944 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453944 Supplier Thermo Scientific Alfa Aesar Supplier No. A11945.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Fluorophenol, 99%, C6H5FO, CAS Number-371-41-5, ccris 665, fpn, 4-fluoro-phenol, p-fluorophenol, phenol, p-fluoro, para-fluorophenol, phenol, 4-fluoro, 4-fluoro phenol, 4-fluoranylphenol, 500g, 1362752, 185 deg.C to 188 deg.C, 1.22, Hazard Class: 6.1; Packaging Group: III, 206-736-0 | CAS: 371-41-5 | C6H5FO | 112.10 g/mol

4-Fluorophenol is a fluorinated phenolic compound with various applications as an starting reagent for the synthesis of pharmaceutical goods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenol is a fluorinated phenolic compound with various applications as an starting reagent for the synthesis of pharmaceutical goods.

Solubility
Soluble in water (80 mg/ml at 20°C), methylene dichloride, chloroform and methanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store away from strong bases and oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-41-5
Molecular Formula C6H5FO
Molecular Weight (g/mol) 112.10
MDL Number MFCD00002316
InChI Key RHMPLDJJXGPMEX-UHFFFAOYSA-N
Synonym p-fluorophenol, phenol, 4-fluoro, phenol, p-fluoro, para-fluorophenol, 4-fluoro phenol, 4-fluoro-phenol, ccris 665, 4-fluoranylphenol, fpn
PubChem CID 9732
IUPAC Name 4-fluorophenol
SMILES OC1=CC=C(F)C=C1

Specifications

Melting Point 44°C to 50°C
Density 1.22
Boiling Point 185°C to 188°C
Flash Point 68°C (154°F)
Odor Phenol-like
Quantity 25 g
UN Number UN2923
Beilstein 1362752
Solubility Information Soluble in water (80mg/ml at 20°C),methylene dichloride,chloroform and methanol.
Formula Weight 112.1
Percent Purity 99%
Chemical Name or Material 4-Fluorophenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 4-Fluorophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.