missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxynonenal Mouse, anti-, Clone: 2C1D10, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | 4-Hydroxynonenal |
|---|---|
| Clone | 2C1D10 |
| Concentration | 1000 μg/mL |
| Applications | ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18017025
|
Proteintech
68538-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18088945
|
Proteintech
68538-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 4-Hydroxynonenal antibody from Proteintech is a mouse monoclonal antibody generated with ptg of na 4-Hydroxynonenal. This antibody recognizes 4-hydroxynonenal antigen. The 4-Hydroxynonenal antibody has been validated for the following applications: ELISA analysis.
Spécification
| 4-Hydroxynonenal | |
| 1000 μg/mL | |
| Liquid | |
| 4-HNE, 4-Hydroxy-2-nonenal, 4-Hydroxynonenal, C9H16O2, Cas. 75899-68-2 | |
| Primary |
| 2C1D10 | |
| ELISA | |
| Mouse | |
| IgG2a | |
| Store at -20°C. Stable for one year after shipment. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit