missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-o-phenylenediamine, 95%

Product Code. 15421689
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421689 1 g 1g
15441689 250 mg 250mg
15431689 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421689 Supplier Thermo Scientific Alfa Aesar Supplier No. H64469.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21304-38-1 | C6H7IN2 | 234.04 g/mol

It is employed in the synthesis of N-(Benzimidazol-2-ylmethyl)iminodiacetic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of N-(Benzimidazol-2-ylmethyl)iminodiacetic acids.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from light, acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21304-38-1
Molecular Formula C6H7IN2
Molecular Weight (g/mol) 234.04
MDL Number MFCD08669466
InChI Key FUOSRKZBOIVBOS-UHFFFAOYSA-N
Synonym 1,2-diamino-4-iodobenzene, 4-iodo-1,2-diaminobenzene, 4-iodo-1,2-phenylenediamine, 1,2-benzenediamine, 4-iodo, 4-iodo-1,2-benzenediamine, 4-iodo-benzene-1,2-diamine, ambkkkkk335, 2-amino-5-iodoaniline, 2-amino4-iodophenylamine
PubChem CID 11207049
IUPAC Name 4-iodobenzene-1,2-diamine
SMILES NC1=C(N)C=C(I)C=C1

Specifications

Melting Point 73°C to 77°C
Quantity 1 g
UN Number UN2811
Beilstein 2716284
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 234.04
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Iodo-o-phenylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.