missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzenesulfonyl chloride, 96%

Product Code. 11378425
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378425 5 g 5g
11388425 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378425 Supplier Thermo Scientific Alfa Aesar Supplier No. L10428.06
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 54997-90-9 | C9H11ClO2S | 218.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54997-90-9
Molecular Formula C9H11ClO2S
Molecular Weight (g/mol) 218.70
MDL Number MFCD00041510
InChI Key CETRNHJIXGITKR-UHFFFAOYSA-N
Synonym 4-isopropylbenzenesulfonyl chloride, 4-isopropylbenzene-1-sulfonyl chloride, 4-isopropylbenzenesulfonylchloride, 4-propan-2-yl benzene-1-sulfonyl chloride, 4-isopropyl benzenesulfonyl chloride, 4-isopropyl-benzenesulfonyl chloride, 4-propan-2-yl benzenesulfonyl chloride, 4-isopropylbenzensulfonylchloride, 4-isopropylphenylsulfonyl chloride, p-isopropylbenzenesulfonyl chloride
PubChem CID 2734882
IUPAC Name 4-propan-2-ylbenzenesulfonyl chloride
SMILES CC(C)C1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Density 1.22
Boiling Point 142°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.539
Quantity 5 g
UN Number UN3265
Beilstein 1103862
Sensitivity Moisture sensitive
Formula Weight 218.7
Percent Purity 96%
Chemical Name or Material 4-Isopropylbenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.