missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzophenone, 98+%

Product Code. 11476336
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476336 25 g 25g
11486336 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476336 Supplier Thermo Scientific Alfa Aesar Supplier No. B23123.14
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 611-94-9 | C14H12O2 | 212.248 g/mol

4-Methoxybenzophenone and its analogues showed diverse plant growth-regulating actions such as inhibition of shoot and root growth, induction of chlorosis, and a disturbance in phototropism or geotropism. It is also used as a photo polymerization catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzophenone and its analogues showed diverse plant growth-regulating actions such as inhibition of shoot and root growth, induction of chlorosis, and a disturbance in phototropism or geotropism. It is also used as a photo polymerization catalyst.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-94-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD00008403
InChI Key SWFHGTMLYIBPPA-UHFFFAOYSA-N
Synonym 4-methoxybenzophenone, 4-methoxyphenyl phenyl methanone, p-methoxybenzophenone, benzophenone, 4-methoxy, methanone, 4-methoxyphenyl phenyl, phenyl p-anisyl ketone, 4-benzoylanisole, 4-methoxyphenyl phenylmethanone, unii-i4xj07373m, 4-methoxyphenyl-phenylmethanone
PubChem CID 69146
IUPAC Name (4-methoxyphenyl)-phenylmethanone
SMILES COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 60°C to 63°C
Boiling Point 354°C to 356°C
Quantity 25 g
Beilstein 1104713
Solubility Information Insoluble in water.
Formula Weight 212.25
Percent Purity ≥98%
Chemical Name or Material 4-Methoxybenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.