missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitrobenzaldehyde, 98%

Product Code. 11313077
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313077 1 g 1g
11323077 5 g 5g
11333077 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11313077 Supplier Thermo Scientific Alfa Aesar Supplier No. L13454.03
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 31680-07-6 | C8H7NO3 | 165.15 g/mol

4-Methyl-3-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-3-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31680-07-6
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00017011
InChI Key KHWGAWBXQOKXIJ-UHFFFAOYSA-N
Synonym 3-nitro-p-tolualdehyde, 4-methyl-3-nitro-benzaldehyde, 3-nitro-4-methylbenzaldehyde, benzaldehyde, 4-methyl-3-nitro, methylnitrobenzaldehyde, pubchem8157, acmc-209hod, 4-methyl-3-nitrobenzaldehyde
PubChem CID 1798998
IUPAC Name 4-methyl-3-nitrobenzaldehyde
SMILES CC1=CC=C(C=O)C=C1[N+]([O-])=O

Specifications

Melting Point 44°C to 48°C
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 1637706
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 165.15
Percent Purity 98%
Chemical Name or Material 4-Methyl-3-nitrobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.