missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzenesulphonic Acid Hydrate, Mikromol™

Product Code. 17242894
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17242894 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17242894 Supplier Mikromol Supplier No. MM0598.02
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Methylbenzenesulphonic Acid Hydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylbenzenesulphonic Acid Hydrate
CAS 6192-52-5
Analyte or Component Names 4-Methylbenzenesulphonic Acid Hydrate
Shipping Condition Room Temperature
Molecular Formula C7 H8 O3 S . H2 O
Certifications/Compliance ISO 17025
Synonym Lisinopril Dihydrate Imp. B (EP), Lisinopril Imp. B (EP), 4-Methylbenzene-sulphonic Acid Hydrate, Lisinopril Dihydrate Imp. B (EP) as Hydrate, Anastrozole Imp. F (EP) as Hydrate, Anastrozole Impurity F as Hydrate, Escitalopram Oxalate Impurity J as Hydrate, Lisinopril Dihydrate Impurity B as Hydrate, Sultamicillin Impurity B as Hydrate, Tizanidine Hydrochloride Impurity I as Hydrate, Escitalopram Impurity J as Hydrate, Lisinopril Impurity B as Hydrate, Tizanidine Impurity I as Hydrate
InChI Formula InChI=1S/C7H8O3S.H2O/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2
SMILES O.Cc1ccc(cc1)S(=O)(=O)O
IUPAC Name 4-methylbenzenesulfonic acid;hydrate
Molecular Weight (g/mol) 190.22
Quantity 100 mg
Formula Weight 190.03 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.