missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylestra-1,3,5(10)-triene-3,17beta-diol (4-Methylestradiol), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
4-Methylestra-1,3,5(10)-triene-3,17beta-diol (4-Methylestradiol)
Specifications
Specifications
| Chemical Name or Material | 4-Methylestradiol |
| CAS | 6171-48-8 |
| Analyte or Component Names | 4-Methylestradiol |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C19 H26 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-Methylestra-1,3,5(10)-triene-3,17beta-diol, Estradiol Hemihydrate Imp. C (EP), 4-Methylestradiol, Estradiol Hemihydrate Impurity C, Estradiol Impurity C |
| InChI Formula | InChI=1S/C19H26O2/c1-11-12-3-4-15-14(13(12)5-7-17(11)20)9-10-19(2)16(15)6-8-18(19)21/h5,7,14-16,18,20-21H,3-4,6,8-10H2,1-2H3/t14-,15-,16+,18+,19+/m1/s1 |
| SMILES | Cc1c(O)ccc2[C@H]3CC[C@]4(C)[C@@H](O)CC[C@H]4[C@@H]3CCc12 |
| IUPAC Name | (8R,9S,13S,14S,17S)-4,13-dimethyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction