missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propoxyphenol, 98%

Product Code. 11405926
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405926 25 g 25g
11495916 5 g 5g
11415926 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405926 Supplier Thermo Scientific Alfa Aesar Supplier No. B22266.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 18979-50-5 | C9H12O2 | 152.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18979-50-5
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00002335
InChI Key KIIIPQXXLVCCQP-UHFFFAOYSA-N
Synonym phenol, 4-propoxy, 4-n-propoxyphenol, p-propoxyphenol, phenol, p-propoxy, hydroquinone monopropyl ether, p-propoxy phenol, 4-propyloxyphenol, 4-propoxy-phenol, pubchem13204, para-propoxyphenol
PubChem CID 29352
IUPAC Name 4-propoxyphenol
SMILES CCCOC1=CC=C(C=C1)O

Specifications

Melting Point 55°C to 58°C
Flash Point >110°C (230°F)
Quantity 25 g
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 4-n-Propoxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.