missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenol, LoGiCal

Product Code. 30460407
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460407 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460407 Supplier LoGiCal Supplier No. LGCFOR0042.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material 4-Nitrophenol
CAS 100-02-7
Analyte or Component Names 4-Nitrophenol
Shipping Condition Room Temperature
Molecular Formula C6 H5 N O3
Certifications/Compliance ISO 9001
Synonym Paracetamol Imp. F (EP), Acetaminophen USP Related Compound F, Acetaminophen USP RC F, 4-Nitrophenol, Ph Eur Paracetamol Impurity F, SKF-30569, p-nitrophenol, Acetaminophen Related Compound F, Paracetamol Impurity F
InChI Formula InChI=1S/C6H5NO3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H
Solution Type Neat
SMILES Oc1ccc(cc1)[N+](=O)[O-]
IUPAC Name 4-nitrophenol
Industry Type Clinical Testing
Molecular Weight (g/mol) 139.11
Quantity 10 mg
Formula Weight 139.0269 g/mol
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.