missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenyl Thiazol-5-ylmethyl Carbonate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
4-Nitrophenyl Thiazol-5-ylmethyl Carbonate
Specifications
Specifications
| Chemical Name or Material | Thiazolylmethyl-4-nitrophenylcarbonate |
| CAS | 144163-97-3 |
| Analyte or Component Names | Thiazolylmethyl-4-nitrophenylcarbonate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H8 N2 O5 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Carbonic acid, 4-nitrophenyl 5-thiazolylmethyl ester, (4-Nitrophenyl) thiazol-5-ylmethyl carbonate, Thiazolylmethyl-4-nitrophenylcarbonate |
| InChI Formula | InChI=1S/C11H8N2O5S/c14-11(17-6-10-5-12-7-19-10)18-9-3-1-8(2-4-9)13(15)16/h1-5,7H,6H2 |
| SMILES | [O-][N+](=O)c1ccc(OC(=O)OCc2cncs2)cc1 |
| IUPAC Name | (4-nitrophenyl) 1,3-thiazol-5-ylmethyl carbonate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction