missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenylacetone, 98%

Product Code. 11464677
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11464677

Brand: Thermo Scientific Alfa Aesar B21183.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5332-96-7 | C9H9NO3 | 179.175 g/mol

It is applied as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an intermediate in organic synthesis.

Solubility
Insoluble in water, soluble in usual organic solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5332-96-7
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00051518
InChI Key GEWWCWZGHNIUBW-UHFFFAOYSA-N
Synonym 4-nitrophenylacetone, 1-4-nitrophenyl propan-2-one, 2-propanone, 1-4-nitrophenyl, 1-4-nitrophenyl-2-propanone, 1-4-nitrophenyl acetone, p-nitrophenylazeton, p-nitrophenylacetone, pubchem18646, 4-nitrophenyl acetone, acmc-1az6k
PubChem CID 219367
IUPAC Name 1-(4-nitrophenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 61°C to 64°C
Quantity 1 g
Beilstein 1953054
Solubility Information Insoluble in water,soluble in usual organic solvents.
Formula Weight 179.18
Percent Purity 98%
Chemical Name or Material 4-Nitrophenylacetone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.