missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrothiophenol, 96%

Product Code. 15465488
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465488 25 g 25g
15455488 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465488 Supplier Thermo Scientific Alfa Aesar Supplier No. H59240.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1849-36-1 | C6H5NO2S | 155.171 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Partly soluble in water and chloroform.

Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1849-36-1
Molecular Formula C6H5NO2S
Molecular Weight (g/mol) 155.171
MDL Number MFCD00007343
InChI Key AXBVSRMHOPMXBA-UHFFFAOYSA-N
Synonym 4-nitrothiophenol, 4-nitrobenzenethiol, p-nitrothiophenol, benzenethiol, 4-nitro, p-nitrophenyl mercaptan, p-nitrobenzenethiol, benzenethiol, p-nitro, 4-nitrobenzene-1-thiol, 4-nitrothiophenolate, 4-nitro-benzenethiol
PubChem CID 15809
SMILES C1=CC(=CC=C1[N+](=O)[O-])S

Specifications

Melting Point 72°C to 77°C
Quantity 25 g
Beilstein 606924
Solubility Information Partly soluble in water and chloroform.
Formula Weight 155.17
Percent Purity 80%
Grade Technical
Chemical Name or Material 4-Nitrothiophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.