missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfobenzoic acid monopotassium salt, 95%

Product Code. 11484017
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484017 50 g 50g
11494017 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484017 Supplier Thermo Scientific Alfa Aesar Supplier No. A19432.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5399-63-3 | C7H5KO5S | 240.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5399-63-3
Molecular Formula C7H5KO5S
Molecular Weight (g/mol) 240.27
MDL Number MFCD00007509
InChI Key PXRJBUPXKDXDLG-UHFFFAOYSA-M
Synonym benzoic acid, 4-sulfo-, monopotassium salt, 4-sulfobenzoic acid monopotassium salt, 4-sulphobenzoic acid monopotassium salt, 4-sulfobenzoicacidmonopotassiumsalt, potassium hydrogen 4-sulphonatobenzoate, p-sulphobenzoic acid, monopotassium salt, potassium 4-sulphobenzoate, benzoic acid, 4-sulfo-, potassium salt 1:1, potassium 4-sulfobenzoate, p-potassiooxysulfonyl benzoic acid
PubChem CID 23670833
IUPAC Name potassium;4-sulfobenzoate
SMILES C1=CC(=CC=C1C(=O)[O-])S(=O)(=O)O.[K+]

Specifications

Quantity 50 g
Sensitivity Hygroscopic
Formula Weight 240.28
Percent Purity 95%
Chemical Name or Material 4-Sulfobenzoic acid monopotassium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.