missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)phenyl isocyanate, 97%, Thermo Scientific™

Product Code. 10049551
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10049551 1 g Amber Glass Bottle 1g
10079571 10 g Amber Glass Bottle 10g
10051144 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10049551 Supplier Thermo Scientific Maybridge Supplier No. SB01802DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1943-67-5
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00040718
InChI Key WUWBDQJTQTVBSQ-UHFFFAOYSA-N
Synonym 4-tert-butylphenyl isocyanate, 4-tert-butylphenylisocyanate, benzene, 1-1,1-dimethylethyl-4-isocyanato, 4-tert-butylphenyl-isocyanate, 4-tert-butyl phenyl isocyanate, 1-tert-butyl-4-isocyanato-benzene, benzene,1-1,1-dimethylethyl-4-isocyanato, 4-t-butylphenylisocyanate, acmc-209ey5, 4-tertbutylphenyl isocyanate
PubChem CID 2735695
IUPAC Name 1-tert-butyl-4-isocyanatobenzene
SMILES CC(C)(C)C1=CC=C(C=C1)N=C=O

Specifications

CAS Min % 97.0
Density 0.986g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)3CC6H4NCO
Quantity 1 g
Formula Weight 175.23
Percent Purity 97%
Chemical Name or Material 4-(Tert-butyl)phenyl isocyanate
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.