missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzaldehyde, 98%

Product Code. 10256740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10256740 50 g Glass bottle 50g
10470671 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10256740 Supplier Thermo Scientific Acros Supplier No. 208920500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 455-19-6 | C8H5F3O | 174.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-19-6
Molecular Formula C8H5F3O
Molecular Weight (g/mol) 174.12
MDL Number MFCD00006952
InChI Key BEOBZEOPTQQELP-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzaldehyde, benzaldehyde, 4-trifluoromethyl, p-trifluoromethylbenzaldehyde, p-trifluoromethyl benzaldehyde, alpha,alpha,alpha-trifluoro-p-tolualdehyde, 4'-trifluoromethyl benzaldehyde, p-trifluoromethyl-benzaldhyde, 4-trifluoromethyl-benzaldehyde
PubChem CID 67996
IUPAC Name 4-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=CC=C1C=O)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2700g/mL
Boiling Point 66°C to 67°C (13.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3C6H4CHO
Refractive Index 1.4625 to 1.4645
Quantity 50 g
Beilstein 07, III, 1020
Specific Gravity 1.27
Formula Weight 174.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethyl)benzaldehyde, 95%
Show More Show Less
compliance-icons
Product Identifier
  • a,a,a-Trifluoro-p-tolualdehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.