missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzhydrol, 97%

Product Code. 11309005
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309005 1 g 1g
11319005 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309005 Supplier Thermo Scientific Alfa Aesar Supplier No. L14147.03
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 395-23-3 | C14H11F3O | 252.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 395-23-3
Molecular Formula C14H11F3O
Molecular Weight (g/mol) 252.236
MDL Number MFCD00014399
InChI Key LIZDGCXCDJOWBS-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzhydrol, phenyl 4-trifluoromethyl phenyl methanol, benzhydrol, 4-trifluoromethyl-,, phenyl-4-trifluoromethyl phenyl methanol, phenyl 4-trifluoromethyl phenyl methan-1-ol, phenyl-4-trifluoromethyl-phenyl, acmc-20amtp, 4-trifluoromethylbenzhydrol, 4-trifluoromethylphenyl phenyl methanol
PubChem CID 258897
IUPAC Name phenyl-[4-(trifluoromethyl)phenyl]methanol
SMILES C1=CC=C(C=C1)C(C2=CC=C(C=C2)C(F)(F)F)O

Specifications

Melting Point 59°C to 61°C
Boiling Point 122°C to 124°C (0.4 mmHg)
Quantity 1 g
Beilstein 2127184
Formula Weight 252.23
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)benzhydrol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.