missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-[(1RS)-1-Methylbutyl]-2-thioxo-2,3-dihydropyrimidine-4,6(1H,5H)-dione, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
5-[(1RS)-1-Methylbutyl]-2-thioxo-2,3-dihydropyrimidine-4,6(1H,5H)-dione
Specifications
Specifications
| Chemical Name or Material | 5-[(1RS)-1-Methylbutyl]-2-thioxo-2,3-dihydropyrimidine-4,6(1H,5H)-dione |
| CAS | 29639-67-6 |
| Analyte or Component Names | 5-[(1RS)-1-Methylbutyl]-2-thioxo-2,3-dihydropyrimidine-4,6(1H,5H)-dione |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H14 N2 O2 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Dihydro-5-(1-methylbutyl)-2-thioxo-4,6(1H,5H)-pyrimidinedione, 5-(1-Methylbutyl)-2-thiobarbituric acid, Thiopental Sodium and Sodium Carbonate Imp. A (EP), Thiopental Imp. A (EP), Thiopental Sodium and Sodium Carbonate Impurity A |
| InChI Formula | InChI=1S/C9H14N2O2S/c1-3-4-5(2)6-7(12)10-9(14)11-8(6)13/h5-6H,3-4H2,1-2H3,(H2,10,11,12,13,14) |
| SMILES | CCCC(C)C1C(=O)NC(=S)NC1=O |
| IUPAC Name | 5-pentan-2-yl-2-sulfanylidene-1,3-diazinane-4,6-dione |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction