missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Hydroxybenzoyl)-4-phenylpentan-2-one, Mikromol™

Product Code. 17921566
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17921566 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17921566 Supplier Mikromol Supplier No. MM0638.020100
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-(2-Hydroxybenzoyl)-4-phenylpentan-2-one
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-(2-Hydroxybenzoyl)-4-phenylpentan-2-one
CAS 2514748-07-1
Analyte or Component Names 5-(2-Hydroxybenzoyl)-4-phenylpentan-2-one
Shipping Condition Ice pack (-20°C)
Molecular Formula C18 H18 O3
Certifications/Compliance ISO 9001
Synonym 5-(2-Hydroxybenzoyl)-4-phenylpentan-2-one, 1-(2-Hydroxyphenyl)-3-phenylhexane-1,5-dione
InChI Formula InChI=1S/C18H18O3/c1-13(19)11-15(14-7-3-2-4-8-14)12-18(21)16-9-5-6-10-17(16)20/h2-10,15,20H,11-12H2,1H3
SMILES CC(=O)CC(CC(=O)c1ccccc1O)c2ccccc2
IUPAC Name 1-(2-hydroxyphenyl)-3-phenylhexane-1,5-dione
Molecular Weight (g/mol) 282.334
Quantity 100 mg
Formula Weight 282.126 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.