missing translation for 'onlineSavingsMsg'
Learn More

DYKDDDDK Epitope Tag Antibody (FG4R), mFluor Violet 610 SE, R&D Systems™

Product Code. 18608279 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18608279 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18608279 Supplier Novus Biologicals Supplier No. NBP237823MFV610
  • ONLINE EXCLUSIVE PRICE
    699.80 €
    602.70 € / 0.1mL
    Save 97.10 €
    13% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

DYKDDDDK Epitope Tag Monoclonal specifically detects DYKDDDDK Epitope Tag in Tag samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYKDDDDK Epitope Tag
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone FG4R
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag
Host Species Mouse
Immunogen DYKDDDDK Epitope Tag Antibody (FG4R) was made to a Synthetic peptide (DYKDDDDK) coupled to KLH.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.