missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydro-4-hydroxy-6-methyl-4H-thieno[2,3-b]thiopyran-2-sulfonamide 7,7-Dioxide, Mikromol™

Product Code. 17903776
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
17903776 25 mg 25mg
1 options
This item is not returnable. View return policy

Product Code. 17903776

Brand: Mikromol MM0403.180025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5,6-Dihydro-4-hydroxy-6-methyl-4H-thieno[2,3-b]thiopyran-2-sulfonamide 7,7-Dioxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5,6-Dihydro-4-hydroxy-6-methyl-4H-thieno[2,3-b]thiopyran-2-sulfonamide 7,7-Dioxide
CAS 120279-26-7
Analyte or Component Names 5,6-Dihydro-4-hydroxy-6-methyl-4H-thieno[2,3-b]thiopyran-2-sulfonamide 7,7-Dioxide
Shipping Condition Room Temperature
Molecular Formula C8H11NO5S3
Certifications/Compliance ISO 9001
Synonym 4H-Thieno[2,3-b]thiopyran-2-sulfonamide, 5,6-dihydro-4-hydroxy-6-methyl-, 7,7-dioxide, 4-Hydroxy-6-methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulfonamide
InChI Formula InChI=1S/C8H11NO5S3/c1-4-2-6(10)5-3-7(17(9,13)14)15-8(5)16(4,11)12/h3-4,6,10H,2H2,1H3,(H2,9,13,14)
SMILES CC1CC(O)c2cc(sc2S1(=O)=O)S(=O)(=O)N
IUPAC Name 4-hydroxy-6-methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulfonamide
Molecular Weight (g/mol) 297.37
Quantity 25 mg
Formula Weight 296.9799 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.