missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-1H-imidazole-4-carboxamide, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
5-Amino-1H-imidazole-4-carboxamide
Specifications
Specifications
| Chemical Name or Material | 5-Amino-1H-imidazole-4-carboxamide |
| CAS | 360-97-4 |
| Analyte or Component Names | 5-Amino-1H-imidazole-4-carboxamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C4 H6 N4 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Dacarbazine Imp. B (EP), Dacarbazine USP Related Compound A, Dacarbazine USP RC A, 5-Amino-1H-imidazole-4-carboxamide, 4-Amino-5-carbamoylimidazole, 4-Aminoimidazole-5-carboxamide, 4-Carbamoyl-5-aminoimidazole, 4-Carboxamido-5-aminoimidazole, 5-Amino-4-carbamoylimidazole, 5-Aminoimidazole-4-carboxamide, AIC, AICA, Ba 2756, NSC 7784, Temozolomide Imp A (EP), Dacarbazine Related Compound A, Dacarbazine Impurity B |
| InChI Formula | InChI=1S/C4H6N4O/c5-3-2(4(6)9)7-1-8-3/h1H,5H2,(H2,6,9)(H,7,8) |
| SMILES | NC(=O)c1nc[nH]c1N |
| IUPAC Name | 5-amino-1H-imidazole-4-carboxamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction