missing translation for 'onlineSavingsMsg'
Learn More

5-Aminonaphthalene-1-sulfonic acid, tech. 85%

Product Code. 11480283
Click to view available options
Quantity:
25 g
50 g
100 g
missing translation for 'unitSize'
100g
25g
50g
Questo articolo non è restituibile. Consulta la politica di reso

Codice prodotto. 11480283

missing translation for 'mfr': Thermo Scientific Alfa Aesar A11433.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Questo articolo non è restituibile. Consulta la politica di reso

CAS: 84-89-9 | C10H9NO3S | 223.25 g/mol

5-Aminonaphthalene-1-sulfonic acid is an deriva- tizing agents in the precolumn derivatization of some chiral phenoxy acid. Cu(II) based on a dansyl derivative was synthesized with 29.2% overall yield by a seven-step synthetic procedure from the readily available starting material 5-aminonaphthalene-1-sulfonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Aminonaphthalene-1-sulfonic acid is an deriva- tizing agents in the precolumn derivatization of some chiral phenoxy acid. Cu(II) based on a dansyl derivative was synthesized with 29.2% overall yield by a seven-step synthetic procedure from the readily available starting material 5-aminonaphthalene-1-sulfonic acid.

Solubility
Soluble in hot water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 84-89-9
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.25
MDL Number MFCD00014315
InChI Key DQNAQOYOSRJXFZ-UHFFFAOYSA-N
Synonym 5-amino-1-naphthalenesulfonic acid, 1-naphthylamine-5-sulfonic acid, laurent acid, 1-naphthalenesulfonic acid, 5-amino, 1-amino-5-sulfonaphthalene, laurent's acid, 5-naphthylamine-1-sulfonic acid, 1-amino-5-naphthalenesufonic acid, 1-aminonaphthalene-5-sulfonic acid, unii-w70wc365od
PubChem CID 6793
IUPAC Name 5-aminonaphthalene-1-sulfonic acid
SMILES NC1=C2C=CC=C(C2=CC=C1)S(O)(=O)=O

Specifica

Melting Point >300°C
Odor Odorless
Quantity 25 g
Beilstein 2214149
Merck Index 14,6404
Solubility Information Soluble in hot water.
Formula Weight 223.25
Percent Purity 90%
Grade Technical
Chemical Name or Material 5-Aminonaphthalene-1-sulfonic acid

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato