missing translation for 'onlineSavingsMsg'
Learn More

5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Ethyl Ester, Mikromol™

Product Code. 30051630
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051630 100 mg 100mg
30052707 25 mg 25mg
30462637 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30051630 Supplier Mikromol Supplier No. MM0154.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Ethyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Ethyl Ester
CAS 3543-74-6
Analyte or Component Names 5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C18 H27 N3 O4
Certifications/Compliance ISO 17025
Synonym 5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Ethyl Ester, [1-Methyl-5-bis(2'-hydroxyethyl)aminobenzimidazolyl-2]butanoic Acid Ethyl Ester, 4-[5-[Bis(2-hydroxyethyl)amino]-1-methylbenzimidazol-2-yl]butanoic acid ethyl ester, Ethyl 5-[bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoate, Bendamustine Related Compound C
InChI Formula InChI=1S/C18H27N3O4/c1-3-25-18(24)6-4-5-17-19-15-13-14(7-8-16(15)20(17)2)21(9-11-22)10-12-23/h7-8,13,22-23H,3-6,9-12H2,1-2H3
SMILES CCOC(=O)CCCc1nc2cc(ccc2n1C)N(CCO)CCO
IUPAC Name ethyl 4-[5-[bis(2-hydroxyethyl)amino]-1-methylbenzimidazol-2-yl]butanoate
Molecular Weight (g/mol) 349.42
Quantity 100 mg
Formula Weight 349.2002 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.