missing translation for 'onlineSavingsMsg'
Learn More

5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Ethyl Ester, Mikromol™

Product Code. 30462637
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462637 4 x 25 mg -
30051630 100 mg 100mg
30052707 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30462637 Supplier Mikromol Supplier No. MM0154.0200254AMP
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Ethyl Ester, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Ethyl Ester
CAS 3543-74-6
Analyte or Component Names 5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C18 H27 N3 O4
Certifications/Compliance ISO 17025
Synonym 5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Ethyl Ester, [1-Methyl-5-bis(2'-hydroxyethyl)aminobenzimidazolyl-2]butanoic Acid Ethyl Ester, 4-[5-[Bis(2-hydroxyethyl)amino]-1-methylbenzimidazol-2-yl]butanoic acid ethyl ester, Ethyl 5-[bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoate, Bendamustine Related Compound C
InChI Formula InChI=1S/C18H27N3O4/c1-3-25-18(24)6-4-5-17-19-15-13-14(7-8-16(15)20(17)2)21(9-11-22)10-12-23/h7-8,13,22-23H,3-6,9-12H2,1-2H3
Solution Type Neat
SMILES CCOC(=O)CCCc1nc2cc(ccc2n1C)N(CCO)CCO
IUPAC Name ethyl 4-[5-[bis(2-hydroxyethyl)amino]-1-methylbenzimidazol-2-yl]butanoate
Molecular Weight (g/mol) 349.42
Quantity 4 x 25 mg
Formula Weight 349.2002 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.