missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,3-difluoroanisole, 97%, Thermo Scientific™

Product Code. 11400058
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400058 2 g 2g
11410058 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400058 Supplier Thermo Scientific Alfa Aesar Supplier No. B23655.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261762-35-0
Molecular Formula C7H5BrF2O
Molecular Weight (g/mol) 223.017
MDL Number MFCD01631352
InChI Key UDTVFZBQFYOZST-UHFFFAOYSA-N
Synonym 5-bromo-2,3-difluoroanisole, 3,4-difluoro-5-methoxy bromobenzene, 2,3-difluoro-5-bromoanisole, benzene, 5-bromo-1,2-difluoro-3-methoxy, 2.3-difluoro-5-bromoanisole, 5-bromo-1,2-difluoro-3-methoxy-benzene, pubchem14832, acmc-1cc5d, 5-bromo-2,3-difluoro anisole
PubChem CID 2773288
IUPAC Name 5-bromo-1,2-difluoro-3-methoxybenzene
SMILES COC1=CC(=CC(=C1F)F)Br

Specifications

Melting Point 33°C to 36°C
Quantity 2 g
Formula Weight 223.02
Percent Purity 97%
Chemical Name or Material 5-Bromo-2,3-difluoroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.