missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
5-Carboxylcytosine Monoclonal antibody specifically detects 5-Carboxylcytosine in Human samples. It is validated for ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair)
Specifications
Specifications
| Antigen | 5-Carboxylcytosine |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Dilution | ELISA, ELISA Capture (Matched Antibody Pair) 50 - 200ng/well, ELISA Detection (Matched Antibody Pair) 0.1 - 0.5ug/ml |
| Gene Alias | 5-caC |
| Host Species | Rabbit |
| Quantity | 1 Set |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?