missing translation for 'onlineSavingsMsg'
Learn More

[5-Chloro-2-(methylamino)phenyl]-phenylmethanone (5-Chloro-2-(methylamino)benzophenone), Mikromol™

Product Code. 30051570
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051570 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051570 Supplier Mikromol Supplier No. MM0023.010025
 Restricted Item
Estimated Shipment: 27-11-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

[5-Chloro-2-(methylamino)phenyl]-phenylmethanone (5-Chloro-2-(methylamino)benzophenone)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloro-2-Methylaminobenzophenone
CAS 1022-13-5
Analyte or Component Names 5-Chloro-2-Methylaminobenzophenone
Shipping Condition Room Temperature
Molecular Formula C14 H12 Cl N O
Certifications/Compliance ISO 17025
Synonym Diazepam Imp. D (EP), Temazepam Imp. A (EP), Diazepam USP Related Compound A, Diazepam USP RC A, [5-Chloro-2-(methylamino)phenyl]phenylmethanone, 5-Chloro-2-(methylamino)benzophenone, Diazepam Related Compound A, Temazepam Related Compound A, Diazepam Impurity D, Temazepam Impurity A
InChI Formula InChI=1S/C14H12ClNO/c1-16-13-8-7-11(15)9-12(13)14(17)10-5-3-2-4-6-10/h2-9,16H,1H3
SMILES CNc1ccc(Cl)cc1C(=O)c2ccccc2
IUPAC Name [5-chloro-2-(methylamino)phenyl]-phenylmethanone
Molecular Weight (g/mol) 245.7
Quantity 25 mg
Formula Weight 245.0607 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.