missing translation for 'onlineSavingsMsg'
Learn More

5-Ethyl-5-(1-ethylpropyl)-2-thioxo-2,3-dihydropyrimidine-4,6(1H,5H)-dione, Mikromol™

Product Code. 17903766
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17903766 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17903766 Supplier Mikromol Supplier No. MM1312.03
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-Ethyl-5-(1-ethylpropyl)-2-thioxo-2,3-dihydropyrimidine-4,6(1H,5H)-dione
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Ethyl-5-(1-ethylpropyl)-2-thioxo-2,3-dihydropyrimidine-4,6(1H,5H)-dione
CAS 87171-21-9
Analyte or Component Names 5-Ethyl-5-(1-ethylpropyl)-2-thioxo-2,3-dihydropyrimidine-4,6(1H,5H)-dione
Shipping Condition Room Temperature
Molecular Formula C11 H18 N2 O2 S
Certifications/Compliance ISO 9001
Synonym Thiopental Sodium and Sodium Carbonate Imp. C (EP), Thiopental Imp. C (EP), 5-Ethyl-5-(1-ethylpropyl)-2-thioxo-2,3-dihydropyrimidine-4,6(1H,5H)-dione, 5-Ethyl-5-(1-ethylpropyl)dihydro-2-thioxo-4,6(1H,5H)-pyrimidinedione, 5-Ethyl-5-(1-ethylpropyl)-2-thiobarbituric acid, Thiopental Sodium and Sodium Carbonate Impurity C
InChI Formula InChI=1S/C11H18N2O2S/c1-4-7(5-2)11(6-3)8(14)12-10(16)13-9(11)15/h7H,4-6H2,1-3H3,(H2,12,13,14,15,16)
SMILES CCC(CC)C1(CC)C(=O)NC(=S)NC1=O
IUPAC Name 5-ethyl-5-pentan-3-yl-2-sulfanylidene-1,3-diazinane-4,6-dione
Molecular Weight (g/mol) 242.34
Quantity 50 mg
Formula Weight 242.1089 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.