missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-HT3E Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 624.00 €
Specifications
| Antigen | 5-HT3E |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18478841
|
Novus Biologicals
NBP2-33578-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18159814
|
Novus Biologicals
NBP2-33578 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-HT3E Polyclonal specifically detects 5-HT3E in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| 5-HT3E | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| A5X5Y0 | |
| 285242 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LAFILSRATPRPALGPLSYREHRVALLHLTHSMSTTGRGVTFTINCSGF | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5-HT3 receptor subunit E splice variant HTR3Ea, 5-HT3c1,5-HT3E, 5-hydroxytryptamine (serotonin) receptor 3, family member E, 5-hydroxytryptamine receptor 3E, MGC120035, MGC120036, serotonin receptor 3E | |
| HTR3E | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title