missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindol-3-acetic Acid, LoGiCal

Product Code. 30460906
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30460906

Brand: LoGiCal LGCFOR0614.11

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

5-Hydroxyindol-3-acetic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Hydroxyindolyl-3-Acetic Acid
CAS 54-16-0
Analyte or Component Names 5-Hydroxyindolyl-3-Acetic Acid
Shipping Condition Room Temperature
Molecular Formula C10 H9 N O3
Certifications/Compliance ISO 9001
Synonym 1 H-Indole-3-acetic acid, 5-hydroxy-, Indole-3-acetic acid, 5-hydroxy- (8 CI), (5-Hydroxy-1 H-indol-3-yl)acetic acid, 2-(5-Hydroxy-1 H-indol-3-yl)acetic acid, 5-HIAA, 5-Hydroxy-1 H-indole-3-acetic acid, 5-Hydroxy-1 H-indole-3-acetic acid, 5-Hydroxy-1 H-indole-3-acetic acid, 5-Hydroxy-1 H-indole-3-acetic acid, 5-Hydroxy-1 H-indole-3-acetic acid, 5-Hydroxy-1 H-indole-3-acetic acid, 5-Hydroxy-1 H-indole-3-acetic acid, 5-Hydroxy-1 H-indole-3-acetic acid, 5-Hydroxy-1 H-indole-3-acetic acid
InChI Formula InChI=1 S/C10H9NO3/c12-7-1-2-9-8(4-7)6(5-11-9)3-10(13)14/h1-2,4-5,11-12 H,3H2,(H,13,14)
SMILES OC(=O)Cc1c[nH]c2ccc(O)cc12
IUPAC Name 2-(5-hydroxy-1 H-indol-3-yl)acetic acid
Industry Type Forensics
Molecular Weight (g/mol) 191.18
Quantity 10 mg
Formula Weight 191.0582 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado